4-Bromophenyl methyl sulfoxide 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-(methylsulphinyl)benzene

A261156-250MG A261156-5G
77238-500EA 15.45 USD
77238-500 77238-502
4-Bromophenyl methyl sulfoxide 98%
4-Bromophenyl methyl sulfoxide

Precaución: for Research Use Only

Formula: C₇H₇BrOS
MW: 219.099 g/mol
MDL Number: MFCD00159074
CAS Number: 934-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR