2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile 95%

Supplier: Ambeed
Warning

A260806-1G A260806-250MG
77262-202EA 120.51 USD
77262-202 77262-204
2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile 95%
2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MW: 186.2100 g/mol
MDL Number: MFCD08166396
CAS Number: 157763-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR