SF1670 98%

Supplier: Ambeed
Warning

Synonyms: N-(9,10-Dioxo-9,10-dihydrophenanthren-2-yl)pivalamide, N-​(9,​10-​dihydro-​9,​10-​dioxo-​2-​phenanthrenyl)-​2,​2-​dimethyl-​propanamide, Protein Tyrosine Phosphatase CD45 Inhibitor, PTPase CD45 Inhibitor

A260724-10MG A260724-1MG A260724-25MG A260724-50MG A260724-5MG
77262-192EA 120.51 USD
77262-192 77262-194 77262-196 77262-198 77262-200
SF1670 98%
SF1670

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₃
MW: 307.34 g/mol
MDL Number: MFCD06411448
CAS Number: 345630-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR