4-Cyano-7-azaindole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Cyano-7-azaindole, 1H-Pyrrolo[2,3-b]pyridine-4-carbonitrile, 7-Azaindole-4-carbonitrile
A260680-10G
A260680-1G
A260680-250MG
A260680-25G
A260680-5G
77289-212EA
230.72
USD
77289-212
77289-214
77289-216
77289-218
77289-220
4-Cyano-7-azaindole 97%
4-Cyano-7-azaindole
Precaución: for Research Use Only
Formula:
C₈H₅N₃ MW: 143.1500 g/mol |
MDL Number:
MFCD08272235 CAS Number: 344327-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...