(2S,3R,4S,5S,6S)-2-(4-(Hydroxymethyl)-2-nitrophenoxy)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate 97%

Supplier: Ambeed
Warning

A260587-100MG A260587-1G A260587-250MG A260587-5G A260587-25G
77250-144EA 30.9 USD
77250-144 77250-146 77250-148 77250-150 77746-428
(2S,3R,4S,5S,6S)-2-(4-(Hydroxymethyl)-2-nitrophenoxy)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate 97%
(2S,3R,4S,5S,6S)-2-(4-(Hydroxymethyl)-2-nitrophenoxy)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO₁₃
MW: 485.39 g/mol
MDL Number: MFCD30489071
CAS Number: 148579-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR