3,5-Difluoroisonicotinaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Difluoro-4-formylpyridine, 3,5-Difluoropyridine-4-carboxaldehyde

A260538-100MG A260538-1G A260538-250MG A260538-5G
77276-536EA 63.86 USD
77276-536 77276-538 77276-540 77276-542
3,5-Difluoroisonicotinaldehyde 95%
3,5-Difluoroisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃F₂NO
MW: 143.091 g/mol
MDL Number: MFCD07368235
CAS Number: 870234-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR