4-Amino-3-mercaptobenzoic acid 97%
Supplier: Ambeed
|
Danger
|
A260063-100MG
A260063-1G
A260063-250MG
A260063-5G
77289-190EA
106.09
USD
77289-190
77289-192
77289-194
77289-196
4-Amino-3-mercaptobenzoic acid 97%
4-Amino-3-mercaptobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₂S MW: 169.20 g/mol |
MDL Number:
MFCD07787433 CAS Number: 14543-45-4 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...