4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine dihydrochloride hydrate 95%

Supplier: Ambeed
Warning

A259996-1G A259996-250MG
77289-186EA 17.51 USD
77289-186 77289-188
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine dihydrochloride hydrate 95%
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine dihydrochloride hydrate

Precaución: for Research Use Only

Formula: C₆H₁₃Cl₂N₃O
MW: 214.09 g/mol
MDL Number: MFCD16875645
CAS Number: 5549-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR