4-Amino-2-methoxy-5-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A259894-100MG A259894-1G A259894-250MG
77238-482EA 40.17 USD
77238-482 77238-484 77238-486
4-Amino-2-methoxy-5-nitrobenzonitrile 97%
4-Amino-2-methoxy-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₇N₃O₃
MW: 193.15 g/mol
MDL Number: MFCD20040384
CAS Number: 1196074-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR