2-Chloro-1,3-benzothiazole-6-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-benzothiazolecarboxylic acid

A259864-250MG A259864-5G
77262-184EA 20.6 USD
77262-184 77262-186
2-Chloro-1,3-benzothiazole-6-carboxylic acid 97%
2-Chloro-1,3-benzothiazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂S
MW: 213.64 g/mol
MDL Number: MFCD08689736
CAS Number: 3855-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR