N-ɑ-Boc-N(im)-tosyl-L-histidine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-1-(p-toluenesulphonyl)-L-histidine, (S)-2-((tert-Butoxycarbonyl)amino)-3-(1-tosyl-1H-imidazol-4-yl)propanoic acid, N-ɑ-(tert-Butoxycarbonyl)-tele-(p-toluenesulphonyl)-L-histidine, Boc-His(Tos)-OH

A259776-10G A259776-1G A259776-25G
77276-526EA 31 USD
77276-526 77276-528 77276-530
N-ɑ-Boc-N(im)-tosyl-L-histidine 98%
N-alpha-Boc-N(im)-tosyl-L-histidine

Precaución: for Research Use Only

Formula: C₁₈H₂₃N₃O₆S
MW: 409.46 g/mol
MDL Number: MFCD00065967
CAS Number: 35899-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR