6'-(Indolin-1-yl)-1,3,3-trimethylspiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine] 95%

Supplier: Ambeed
Warning

A259335-100G A259335-10G A259335-1G A259335-25G A259335-5G
77250-120EA 937.3 USD
77250-120 77250-122 77250-124 77250-126 77250-128
6'-(Indolin-1-yl)-1,3,3-trimethylspiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine] 95%
6'-(Indolin-1-yl)-1,3,3-trimethylspiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine]

Precaución: for Research Use Only

Formula: C₃₀H₂₇N₃O
MW: 445.56 g/mol
MDL Number: MFCD00329019
CAS Number: 114747-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR