2,7-Dibromospiro[9H-fluorene-9,9'-[9H]xanthene] 97%

Supplier: Ambeed
Warning

A259279-10G A259279-1G A259279-250MG A259279-25G
77238-468EA 109 USD
77238-468 77238-470 77238-472 77238-474
2,7-Dibromospiro[9H-fluorene-9,9'-[9H]xanthene] 97%
2,7-Dibromospiro[9H-fluorene-9,9'-[9H]xanthene]

Precaución: for Research Use Only

Formula: C₂₅H₁₄Br₂O
MDL Number: MFCD30342413
CAS Number: 198142-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR