4,4'-Dibromo-4''-methoxytriphenylamine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-N-(4-bromophenyl)-N-(4-methoxyphenyl)aniline, N,N-Bis(4-bromophenyl)-4-methoxyaniline

A259238-1G A259238-250MG A259238-5G
77262-174EA 41 USD
77262-174 77262-176 77262-178
4,4'-Dibromo-4''-methoxytriphenylamine 97%
4,4'-Dibromo-4''-methoxytriphenylamine

Precaución: for Research Use Only

Formula: C₁₉H₁₅Br₂NO
MDL Number: MFCD31699798
CAS Number: 100308-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR