3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3'-Amino-2'-hydroxybiphenyl-3-carboxylic Acid, 3'-Amino-2'-hydroxy-[1,1'-biphenyl]-3-carboxylic acid, 3-(3-Amino-2-hydroxyphenyl)benzoic acid

A259179-100MG A259179-1G A259179-250MG A259179-5G
77276-504EA 15.45 USD
77276-504 77276-506 77276-508 77276-510
3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid 97%
3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
MDL Number: MFCD10001467
CAS Number: 376592-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR