Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II) 98%

Supplier: Ambeed
Warning

Synonyms: Ethanone, 1-(4-chlorophenyl)-2-[5-[2-(1H-tetrazol-1-yl)phenyl]-2H-tetrazol-2-yl]-, Dichlorobis[3-(diphenylphosphino-κP)-1-propanamine-κN]ruthenium, 1-(4-Chloro-phenyl)-2-[5-(2-tetrazol-1-yl-phenyl)-tetrazol-2-yl]-ethanone, 1-(4-Chlorophenyl)-2-{5-[2-(1H-tetrazol-1-yl)phenyl]-2H-tetrazol-2-yl}ethanone

A259162-100MG A259162-1G A259162-250MG
77262-168EA 21 USD
77262-168 77262-170 77262-172
Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II) 98%
Dichlorobis(3-(diphenylphosphino)propylamine)ruthenium(II)

Precaución: for Research Use Only

Formula: C₃₀H₃₆Cl₂N₂P₂Ru
MW: 658.54 g/mol
MDL Number: MFCD15144861
CAS Number: 911128-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR