2-Chloroethyl toluene-4-sulfonate 98%

Supplier: Ambeed
Warning

A259006-1G A259006-5G A259006-100G
77250-116EA 12 USD
77250-116 77250-118 77616-088
2-Chloroethyl toluene-4-sulfonate 98%
2-Chloroethyl toluene-4-sulphonate
Formula: C₉H₁₁ClO₃S
MDL Number: MFCD00000970
CAS Number: 80-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR