2,2'-Bicinchoninic acid dipotassium salt hydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4'-Dicarboxy-2,2'-biquinoline Dipotassium Salt, Potassium [2,2'-biquinoline]-4,4'-dicarboxylate, 2,2'-Biquinoline-4,4'-dicarboxylic Acid Dipotassium Salt
A258913-100G
A258913-25G
A258913-5G
77250-110EA
198.79
USD
77250-110
77250-112
77250-114
2,2'-Bicinchoninic acid dipotassium salt hydrate 98%
2,2'-Bicinchoninic acid dipotassium salt hydrate
Precaución: for Research Use Only
Formula:
C₂₀H₁₂N₂O₄ MW: 420.51 g/mol |
MDL Number:
MFCD00014664 CAS Number: 63451-34-3 |
Specification Test Results
Loss on drying | ≤ 10% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...