(5-Chloro-2-pyridinyl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-(hydroxymethyl)pyridine, (5-Chloropyridin-2-yl)methanol

A258807-1G A258807-25G A258807-100MG A258807-250MG
77276-500EA 44.29 USD
77276-500 77276-502 77748-487 77748-488
(5-Chloro-2-pyridinyl)methanol 95%
(5-Chloro-2-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.5709 g/mol
MDL Number: MFCD10000856
CAS Number: 209526-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR