tert-Butyl 3-bromo-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate 97%

Supplier: Ambeed
Warning

A258753-100MG A258753-250MG
77262-158EA 164.8 USD
77262-158 77262-160
tert-Butyl 3-bromo-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate 97%
tert-Butyl 3-bromo-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN₃O₂
MW: 288.14 g/mol
MDL Number: MFCD30188965
CAS Number: 1784831-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR