1-Amino-1H-pyrrole-2-carbonitrile 95%

Supplier: Ambeed
Danger

A258717-100MG A258717-1G A258717-250MG A258717-5G A258717-10G
77238-448EA 9.27 USD
77238-448 77238-450 77238-452 77238-454 77616-084
1-Amino-1H-pyrrole-2-carbonitrile 95%
1-Amino-1H-pyrrole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₅N₃
MDL Number: MFCD09025773
CAS Number: 159326-66-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR