4,5-Dichloropyridazine-3,6-diol 97%

Supplier: Ambeed
Warning

A258592-10G A258592-1G A258592-5G
77289-154EA 134.93 USD
77289-154 77289-156 77289-158
4,5-Dichloropyridazine-3,6-diol 97%
4,5-Dichloropyridazine-3,6-diol

Precaución: for Research Use Only

Formula: C₄H₂Cl₂N₂O₂
MW: 180.98 g/mol
MDL Number: MFCD00834958
CAS Number: 6641-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR