2,6-Dimethoxybenzonitrile 98%

Supplier: Ambeed
Warning

A258504-10G A258504-1G A258504-25G A258504-5G
77276-480EA 36.05 USD
77276-480 77276-482 77276-484 77276-486
2,6-Dimethoxybenzonitrile 98%
2,6-Dimethoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00001788
CAS Number: 16932-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR