NSC 23005 Sodium 97%

Supplier: Ambeed
Warning

Synonyms: NSC23005 Sodium, Sodium 4-(N-cyclohexylsulphamoyl)benzoate

A258442-10MG A258442-1MG A258442-25MG A258442-50MG A258442-5MG
77262-148EA 28 USD
77262-148 77262-150 77262-152 77262-154 77262-156
NSC 23005 Sodium 97%
NSC 23005 Sodium

Precaución: for Research Use Only

Formula: C₁₃H₁₆NNaO₄S
MDL Number: MFCD30534395
CAS Number: 1796596-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR