4-Methyl-3,5-dinitrobenzoic acid 95%

Supplier: Ambeed
Warning

A258400-100G A258400-10G A258400-25G A258400-500G
77238-440EA 31 USD
77238-440 77238-442 77238-444 77238-446
4-Methyl-3,5-dinitrobenzoic acid 95%
4-Methyl-3,5-dinitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₆
MDL Number: MFCD00007175
CAS Number: 16533-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR