18-Methoxy-18-oxooctadecanoic acid 97%

Supplier: Ambeed
Warning

A258335-100MG A258335-250MG A258335-5G
77262-142EA 26 USD
77262-142 77262-144 77262-146
18-Methoxy-18-oxooctadecanoic acid 97%
18-Methoxy-18-oxooctadecanoic acid

Precaución: for Research Use Only

Formula: C₁₉H₃₆O₄
MDL Number: MFCD00673469
CAS Number: 72849-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR