PD-1/PD-L1 Inhibitor 2 98%

Supplier: Ambeed
Warning

A257865-10MG A257865-1MG A257865-5MG A257865-250MG A257865-100MG A257865-1G A257865-50MG
77238-422EA 43 USD
77238-422 77238-424 77238-426 77616-080 77746-418 77746-419 77746-420
PD-1/PD-L1 Inhibitor 2 98%
PD-1/PD-L1 Inhibitor 2

Precaución: for Research Use Only

Formula: C₂₅H₂₉N₃O₃
MDL Number: MFCD29905455
CAS Number: 1675203-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR