(3aR,8aR)-4,4,8,8-Tetrakis(3,5-diethylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine 98%

Supplier: Ambeed
Warning

A257864-100MG A257864-1G A257864-250MG
77250-098EA 109.18 USD
77250-098 77250-100 77250-102
(3aR,8aR)-4,4,8,8-Tetrakis(3,5-diethylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine 98%
(3aR,8aR)-4,4,8,8-Tetrakis(3,5-diethylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine

Precaución: for Research Use Only

Formula: C₅₃H₆₅O₄P
MW: 797.05 g/mol
MDL Number: MFCD23704801
CAS Number: 1187446-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR