2-(4-Bromonaphthalen-1-yl)-4,6-diphenyl-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A257665-10G A257665-5G
77262-130EA 144.2 USD
77262-130 77262-132
2-(4-Bromonaphthalen-1-yl)-4,6-diphenyl-1,3,5-triazine 97%
2-(4-Bromonaphthalen-1-yl)-4,6-diphenyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₅H₁₆BrN₃
MW: 438.31 g/mol
MDL Number: MFCD30469069
CAS Number: 1800228-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR