4,5-Dimethoxyanthranilic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4,5-dimethoxybenzoic acid, 6-Aminoveratric acid

A257401-1000G A257401-100G A257401-10G A257401-25G A257401-500G A257401-5G
77250-070EA 504.7 USD
77250-070 77250-072 77250-074 77250-076 77250-078 77250-080
4,5-Dimethoxyanthranilic acid 98%
4,5-Dimethoxyanthranilic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
MDL Number: MFCD00011671
CAS Number: 5653-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR