4,4',4'',4'''-([1,1'-Biphenyl]-4,4'-diylbis(azanetriyl))tetrabenzaldehyde 97%

Supplier: Ambeed
Warning

A257325-100MG A257325-250MG
77262-128EA 61.8 USD
77262-128 77748-481
4,4',4'',4'''-([1,1'-Biphenyl]-4,4'-diylbis(azanetriyl))tetrabenzaldehyde 97%
4,4',4'',4'''-([1,1'-Biphenyl]-4,4'-diylbis(azanetriyl))tetrabenzaldehyde

Precaución: for Research Use Only

Formula: C₄₀H₂₈N₂O₄
MW: 600.66 g/mol
MDL Number: MFCD12031339
CAS Number: 865448-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR