4-Amino-m-anisic acid 97%

Supplier: Ambeed
Warning

A257298-100G A257298-10G A257298-1G A257298-25G A257298-5G
77250-060EA 155 USD
77250-060 77250-062 77250-064 77250-066 77250-068
4-Amino-m-anisic acid 97%
4-Amino-m-anisic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD00016539
CAS Number: 2486-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR