2,5-Difluoro-4-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A257024-10G A257024-1G A257024-250MG A257024-5G
77250-052EA 176.13 USD
77250-052 77250-054 77250-056 77250-058
2,5-Difluoro-4-methoxybenzaldehyde 98%
2,5-Difluoro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD06246882
CAS Number: 879093-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR