7-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)-N-hydroxyheptanamide 98%

Supplier: Ambeed
Warning

A256760-10MG A256760-1MG A256760-25MG A256760-50MG A256760-5MG
77238-392EA 291 USD
77238-392 77238-394 77238-396 77238-398 77238-400
7-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)-N-hydroxyheptanamide 98%
7-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)-N-hydroxyheptanamide

Precaución: for Research Use Only

Formula: C₁₉H₂₈Cl₂N₄O₂
MW: 415.36 g/mol
MDL Number: MFCD28400036
CAS Number: 1236199-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR