2-(Boc-amino)-5-bromopyridine 98%

Supplier: Ambeed
Warning

A256579-100G A256579-25G A256579-5G
77250-040EA 215 USD
77250-040 77250-042 77250-044
2-(Boc-amino)-5-bromopyridine 98%
2-(Boc-amino)-5-bromopyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₂
MDL Number: MFCD06411381
CAS Number: 159451-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR