EED-226 98%

Supplier: Ambeed
Warning

Synonyms: N-(Furan-2-ylmethyl)-8-(4-(methylsulphonyl)phenyl)-[1,2,4]triazolo[4,3-c]pyrimidin-5-amine, N-(Furan-2-ylmethyl)-8-(4-(methylsulphonyl)phenyl)[1,2,4]-triazolo[4,3-c]pyrimidin-5-amine, EED226

A256458-10MG A256458-1MG A256458-25MG A256458-5MG
77262-118EA 56 USD
77262-118 77262-120 77262-122 77262-124
EED-226 98%
EED-226

Precaución: for Research Use Only

Formula: C₁₇H₁₅N₅O₃S
MW: 369.4 g/mol
MDL Number: MFCD30738018
CAS Number: 2083627-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR