(9H-Fluoren-9-yl)methyl 3-oxopiperazine-1-carboxylate 98%

Supplier: Ambeed
Danger

A256135-100MG A256135-1G A256135-250MG A256135-5G
77250-028EA 12.36 USD
77250-028 77250-030 77250-032 77250-034
(9H-Fluoren-9-yl)methyl 3-oxopiperazine-1-carboxylate 98%
(9H-Fluoren-9-yl)methyl 3-oxopiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₂O₃
MW: 322.35 g/mol
MDL Number: MFCD02181114
CAS Number: 1119449-40-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR