3-Amino-5-cyanobenzeneboronic acid hydrochloride 97%

Supplier: Ambeed
Warning

A255897-100MG A255897-1G A255897-250MG
77276-424EA 14 USD
77276-424 77276-426 77276-428
3-Amino-5-cyanobenzeneboronic acid hydrochloride 97%
3-Amino-5-cyanobenzeneboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₂·ClH
MDL Number: MFCD08436033
CAS Number: 913835-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR