5-(3-Chlorophenyl)oxazole 98%

Supplier: Ambeed
Warning

A255700-1G A255700-250MG A255700-5G
77262-108EA 116 USD
77262-108 77262-110 77262-112
5-(3-Chlorophenyl)oxazole 98%
5-(3-Chlorophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD00084974
CAS Number: 89808-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR