(E)-6-Bromo-3-(6-bromo-1-(2-octyldodecyl)-2-oxoindolin-3-ylidene)-1-(2-octyldodecyl)indolin-2-one 97%

Supplier: Ambeed
Warning

A255423-100MG A255423-250MG
77250-014EA 33.99 USD
77250-014 77250-016
(E)-6-Bromo-3-(6-bromo-1-(2-octyldodecyl)-2-oxoindolin-3-ylidene)-1-(2-octyldodecyl)indolin-2-one 97%
(E)-6-Bromo-3-(6-bromo-1-(2-octyldodecyl)-2-oxoindolin-3-ylidene)-1-(2-octyldodecyl)indolin-2-one

Precaución: for Research Use Only

Formula: C₅₆H₈₈Br₂N₂O₂
MW: 981.12 g/mol
MDL Number: MFCD30489176
CAS Number: 1375101-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR