3-Chloro-2,4-difluoroaniline 98%

Supplier: Ambeed
Warning

A255420-100G A255420-10G A255420-1G A255420-25G A255420-5G
77276-412EA 183 USD
77276-412 77276-414 77276-416 77276-418 77276-420
3-Chloro-2,4-difluoroaniline 98%
3-Chloro-2,4-difluoroaniline

Precaución: for Research Use Only

Formula: C6H4ClF2N
MDL Number: MFCD00042200
CAS Number: 2613-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR