2-Phenyl-4-thiazolecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylthiazole-4-carboxylic acid

A255287-250MG A255287-25G
77276-408EA 11.33 USD
77276-408 77276-410
2-Phenyl-4-thiazolecarboxylic acid 95%
2-Phenyl-4-thiazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂S
MW: 205.23 g/mol
MDL Number: MFCD00141954
CAS Number: 7113-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR