3-Bromoquinoline 98%

Supplier: Ambeed
Warning

A254837-100G A254837-10G A254837-25G A254837-5G
77262-084EA 19 USD
77262-084 77262-086 77262-088 77262-090
3-Bromoquinoline 98%
3-Bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
MDL Number: MFCD00006767
CAS Number: 5332-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR