N-(2-((S)-2-(5-((S)-3-Aminopyrrolidin-1-yl)-6-methylpyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carbonyl)-4-chlorophenyl)methanesulfonamide 98%

Supplier: Ambeed
Warning

A254733-1MG A254733-5MG
77238-378EA 77 USD
77238-378 77238-380
N-(2-((S)-2-(5-((S)-3-Aminopyrrolidin-1-yl)-6-methylpyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carbonyl)-4-chlorophenyl)methanesulfonamide 98%
N-(2-((S)-2-(5-((S)-3-Aminopyrrolidin-1-yl)-6-methylpyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carbonyl)-4-chlorophenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₂₄H₃₀ClN₇O₃S
MDL Number: MFCD28963961
CAS Number: 1353625-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR