N-Boc-3-ethylpiperazine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Ethylpiperazine, N1-BOC protected, N-Boc-3-Ethylpiperazine

A254701-1G A254701-250MG
77276-392EA 48.41 USD
77276-392 77276-394
N-Boc-3-ethylpiperazine 95%
N-Boc-3-ethylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
MDL Number: MFCD07367735
CAS Number: 438049-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR