5-Aminoindan 98%

Supplier: Ambeed
Warning

A254356-1G A254356-25G A254356-5G
77250-008EA 6 USD
77250-008 77250-010 77250-012
5-Aminoindan 98%
5-Aminoindan

Precaución: for Research Use Only

Formula: C₉H₁₁N
MDL Number: MFCD00003803
CAS Number: 24425-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR