Methyl 2-((tetrahydro-2H-pyran-2-yl)oxy)acetate 97%

Supplier: Ambeed
Warning

A253951-100MG A253951-250MG
77249-994EA 26.78 USD
77249-994 77249-996
Methyl 2-((tetrahydro-2H-pyran-2-yl)oxy)acetate 97%
Methyl 2-((tetrahydro-2H-pyran-2-yl)oxy)acetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
MW: 174.19 g/mol
MDL Number: MFCD18253459
CAS Number: 135643-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR