Bis(((2S,4S)-4-(4-(3-methyl-1-phenyl-1H-pyrazol-5-yl)piperazin-1-yl)pyrrolidin-2-yl)(thiazolidin-3-yl)methanone) pentahydrobromide 98%

Supplier: Ambeed
Warning

Synonyms: Teneligliptin HBr

A253925-10MG A253925-50MG A253925-250MG A253925-1G A253925-1MG
77238-362EA 5.15 USD
77238-362 77238-364 77616-062 77616-058 77616-060
Bis(((2S,4S)-4-(4-(3-methyl-1-phenyl-1H-pyrazol-5-yl)piperazin-1-yl)pyrrolidin-2-yl)(thiazolidin-3-yl)methanone) pentahydrobromide 98%
Bis(((2S,4S)-4-(4-(3-methyl-1-phenyl-1H-pyrazol-5-yl)piperazin-1-yl)pyrrolidin-2-yl)(thiazolidin-3-yl)methanone) pentahydrobromide

Precaución: for Research Use Only

Formula: C₄₄H₆₅Br₅N₁₂O₂S₂
MW: 1257.72 g/mol
MDL Number: MFCD23098790
CAS Number: 906093-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR