3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, (4-Methylpiperazin-1-yl)(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

A253857-1G A253857-250MG A253857-5G
77276-378EA 47 USD
77276-378 77276-380 77276-382
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 98%
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₇BN₂O₃
MW: 330.23 g/mol
MDL Number: MFCD05864308
CAS Number: 883738-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR