1-(2,4,6-Trimethylphenyl)-3-(cyclododecyl)imidazolium chloride 98%

Supplier: Ambeed
Warning

A253840-100MG A253840-1G A253840-250MG
77249-986EA 58 USD
77249-986 77249-988 77249-990
1-(2,4,6-Trimethylphenyl)-3-(cyclododecyl)imidazolium chloride 98%
1-(2,4,6-Trimethylphenyl)-3-(cyclododecyl)imidazolium chloride

Precaución: for Research Use Only

Formula: C₂₄H₃₇ClN₂
MDL Number: MFCD28139676
CAS Number: 1583244-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR